Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-aminophenyl) Sulfide 98.0+%, TCI America™
SDP

Catalog No. T063225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T063225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T063225G Supplier TCI America Supplier No. T063225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 139-65-1
Molecular Formula C12H12N2S
Molecular Weight (g/mol) 216.30
MDL Number MFCD00025342
InChI Key ICNFHJVPAJKPHW-UHFFFAOYSA-N
Synonym 4,4'-thiodianiline, 4,4'-diaminodiphenyl sulfide, bis 4-aminophenyl sulfide, 4,4'-thioaniline, benzenamine, 4,4'-thiobis, p,p-thiodianiline, thiodi-p-phenylenediamine, bis p-aminophenyl sulfide, di p-aminophenyl sulfide
PubChem CID 8765
ChEBI CHEBI:82374
IUPAC Name 4-[(4-aminophenyl)sulfanyl]aniline
SMILES NC1=CC=C(SC2=CC=C(N)C=C2)C=C1

Specifications

Melting Point 108°C
Quantity 25 g
UN Number 3077
Formula Weight 216.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-aminophenyl) Sulfide
EINECSNumber (3)-1110
RTECSNumber BY9625000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.