Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-bromophenyl)phenylphosphine Oxide 98.0+%, TCI America™
SDP

Catalog No. B5532200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5532200MG 200 mg
B55321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5532200MG Supplier TCI America Supplier No. B5532200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93869-52-4
Molecular Formula C18H13Br2OP
Molecular Weight (g/mol) 436.083
InChI Key URGNHDJCYWEAKG-UHFFFAOYSA-N
PubChem CID 15469924
IUPAC Name 1-bromo-4-[(4-bromophenyl)-phenylphosphoryl]benzene
SMILES C1=CC=C(C=C1)P(=O)(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 280°C
Quantity 200 mg
Formula Weight 436.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-bromophenyl)phenylphosphine Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.