Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-chloro-2-nitrophenyl) Disulfide 98.0+%, TCI America™
SDP

Catalog No. B424725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B424725G 25 g
B42475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B424725G Supplier TCI America Supplier No. B424725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2050-66-0
Molecular Formula C12H6Cl2N2O4S2
Molecular Weight (g/mol) 377.21
MDL Number MFCD00047692
InChI Key DESADCWXGJLRSR-UHFFFAOYSA-N
PubChem CID 74907
IUPAC Name 4-chloro-1-[(4-chloro-2-nitrophenyl)disulfanyl]-2-nitrobenzene
SMILES C1=CC(=C(C=C1Cl)[N+](=O)[O-])SSC2=C(C=C(C=C2)Cl)[N+](=O)[O-]

Specifications

Melting Point 216°C
Quantity 25 g
Synonym 4,4′-Dichloro-2,2′-dinitrodiphenyl Disulfide
Formula Weight 377.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-chloro-2-nitrophenyl) Disulfide
EINECSNumber (3)-1131
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.