Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-chlorophenyl) sulfoxide, 98%, Thermo Scientific™

Catalog No. AAA1100422
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1100422 100 g
AAA1100406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1100422 Supplier Thermo Scientific Chemicals Supplier No. A1100422

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3085-42-5
Molecular Formula C12H8Cl2OS
Molecular Weight (g/mol) 271.155
MDL Number MFCD00000618
InChI Key KJGYFISADIZFEL-UHFFFAOYSA-N
Synonym 4-chlorophenyl sulfoxide, bis 4-chlorophenyl sulfoxide, bis p-chlorophenyl sulfoxide, p-chlorophenyl sulfoxide, di-p-chlorophenyl sulfoxide, sulfoxide, bis p-chlorophenyl, 4,4-dichlorodiphenyl sulfoxide, p,p'-dichlorodiphenyl sulfoxide, 4,4'-dichlorodiphenylsulfoxide
PubChem CID 18329
IUPAC Name 1-chloro-4-(4-chlorophenyl)sulfinylbenzene
SMILES C1=CC(=CC=C1S(=O)C2=CC=C(C=C2)Cl)Cl

Specifications

Melting Point 142°C to 144°C
Quantity 100 g
Formula Weight 272.17
Percent Purity 98%
Chemical Name or Material Bis (4-chlorophenyl) sulfoxide
RTECSNumber WS2275000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.