Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-fluorophenyl) Ether 98.0+%, TCI America™
SDP

Catalog No. B26485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26485G 5 g
B26481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26485G Supplier TCI America Supplier No. B26485G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 330-93-8
Molecular Formula C12H8F2O
Molecular Weight (g/mol) 206.19
MDL Number MFCD00013552
InChI Key UUKHFGSOCZLVJO-UHFFFAOYSA-N
Synonym bis 4-fluorophenyl ether, 4,4'-oxybis fluorobenzene, 4-fluorophenyl ether, 4,4'-difluorodiphenyl ether, 1-fluoro-4-4-fluorophenoxy benzene, 1,1'-oxybis 4-fluorobenzene, unii-9r9o553twz, p-fluorophenyl ether, benzene, 1,1'-oxybis 4-fluoro
PubChem CID 67615
IUPAC Name 1-fluoro-4-(4-fluorophenoxy)benzene
SMILES FC1=CC=C(OC2=CC=C(F)C=C2)C=C1

Specifications

Color Yellow
Boiling Point 240°C
Quantity 5 g
Formula Weight 206.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(4-fluorophenyl) Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.