Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-methacryloylthiophenyl) Sulfide 97.0+%, TCI America™
SDP

Catalog No. B166225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B166225G 25 g
B16625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B166225G Supplier TCI America Supplier No. B166225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 129283-82-5
Molecular Formula C20H18O2S3
Molecular Weight (g/mol) 386.54
MDL Number MFCD00142120
InChI Key SPNAQSNLZHHUIJ-UHFFFAOYSA-N

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 25 g
Synonym 4,4′-Thiodibenzenedithiol Dimethacrylate
Formula Weight 386.54
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-methacryloylthiophenyl) Sulfide
EINECSNumber (3)-4098
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.