Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-methylpentyl)phthalate, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013761 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3761 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3761 Supplier MilliporeSigma Supelco Supplier No. 41260100MG
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 259139-51-0
Linear Formula C20H30O4
MDL Number MFCD00072270
Synonym Phthalic acid bis(4-methylpentyl) ester
Type Analytical Standard
Quantity 100 mg
Formula Weight 334.45
Percent Purity ≥95.0% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.