Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-nitrophenyl)amine 98.0+%, TCI America™
SDP

Catalog No. B29605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29605G 5 g
B29601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29605G Supplier TCI America Supplier No. B29605G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1821-27-8
Molecular Formula C12H9N3O4
Molecular Weight (g/mol) 259.221
MDL Number MFCD00437044
InChI Key MTWHRQTUBOTQTE-UHFFFAOYSA-N
PubChem CID 235381
IUPAC Name 4-nitro-N-(4-nitrophenyl)aniline
SMILES C1=CC(=CC=C1NC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 216°C
Quantity 5 g
Synonym 4,4′-Dinitrodiphenylamine
Formula Weight 259.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-nitrophenyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.