Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-nitrophenyl) Carbonate 98.0+%, TCI America™
SDP

Catalog No. C148125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C148125G 25 g
C14815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C148125G Supplier TCI America Supplier No. C148125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5070-13-3
Molecular Formula C13H8N2O7
Molecular Weight (g/mol) 304.214
MDL Number MFCD00007322
InChI Key ACBQROXDOHKANW-UHFFFAOYSA-N
Synonym bis 4-nitrophenyl carbonate, bis p-nitrophenyl carbonate, 4,4'-dinitrodiphenyl carbonate, carbonic acid, bis 4-nitrophenyl ester, p,p'-dinitrodiphenylcarbonate, carbonic acid bis 4-nitrophenyl ester, carbonic acid, bis p-nitrophenyl ester, npc bis 4-nitrophenyl carbonate
PubChem CID 78756
IUPAC Name bis(4-nitrophenyl) carbonate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC(=O)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 25 g
Formula Weight 304.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-nitrophenyl) Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.