Learn More
Medchemexpress LLC Bis(4-nitrophenyl) phosphate | 645-15-8 | 99.9% | 340.18 | 1 G
SDP
Catalog No. 5000454755
Encompass_Preferred
Bis(4-nitrophenyl) phosphate is a catalyst for metal micelles and can catalyze the hydrolysis of (4-nitrophenyl) phosphate. It is also an effective catalyst for ring-opening polymerization (ROP), which can achieve controlled polymerization of β-butyrolactone (β-BL) ring-opening and can be used to prepare diembedded polymers without quenching segment copolymer.
  • For research use only.
Catalog No. 50-004-54755 Supplier Medchemexpress LLC Supplier No. HYW0132051G
Bis(4-nitrophenyl) phosphate is a catalyst for metal micelles and can catalyze the hydrolysis of (4-nitrophenyl) phosphate. It is also an effective catalyst for ring-opening polymerization (ROP), which can achieve controlled polymerization of β-butyrolactone (β-BL) ring-opening and can be used to prepare diembedded polymers without quenching segment copolymer.
  • For research use only.