Learn More
Medchemexpress LLC Bis(4-nitrophenyl) phosphate | 645-15-8 | 99.9% | 340.18 | 500 MG
SDP
Catalog No. 5000454433
Encompass_Preferred
Bis(4-nitrophenyl) phosphate is a catalyst for metal micelles, capable of catalyzing the hydrolysis of (4-nitrophenyl) phosphate. It is also an effective catalyst for ring-opening polymerization (ROP), enabling controlled polymerization of β-butyrolactone (β-BL) ring-opening to prepare diembedded segment copolymer without quenching.
  • Catalyst for metal micelles.
  • Catalyzes the hydrolysis of (4-nitrophenyl) phosphate.
  • Effective catalyst for ring-opening polymerization (ROP).
  • Can achieve controlled polymerization of β-butyrolactone (β-BL) ring-opening.
  • Can be used to prepare diembedded polymers without quenching segment copolymer.
Catalog No. 50-004-54433 Supplier Medchemexpress LLC Supplier No. HYW013205500MG
Bis(4-nitrophenyl) phosphate is a catalyst for metal micelles, capable of catalyzing the hydrolysis of (4-nitrophenyl) phosphate. It is also an effective catalyst for ring-opening polymerization (ROP), enabling controlled polymerization of β-butyrolactone (β-BL) ring-opening to prepare diembedded segment copolymer without quenching.
  • Catalyst for metal micelles.
  • Catalyzes the hydrolysis of (4-nitrophenyl) phosphate.
  • Effective catalyst for ring-opening polymerization (ROP).
  • Can achieve controlled polymerization of β-butyrolactone (β-BL) ring-opening.
  • Can be used to prepare diembedded polymers without quenching segment copolymer.