Learn More
Medchemexpress LLC Bis(4-octylphenyl)amine | 101-67-7 | MFCD00048942 | 393.65 g·mol⁻¹ | C28H43N | 10 MG
SDP
Catalog No. 5000301349
Encompass_Preferred
Bis(4-octylphenyl)amine (CAS 101-67-7) is a nitrogen-containing organic compound used as a high-temperature antioxidant additive in lubricants and studied as an anthropogenic environmental contaminant. Supplied as a research reagent, it has molecular formula C28H43N and molecular weight 393.65 g·mol⁻¹.
  • Used to evaluate antioxidant performance at elevated temperatures.
  • Relevant for environmental contaminant and fate studies.
  • Available in small, research-scale quantities for analytical work.
  • Supplied with molecular identity information (CAS, formula, mass).
  • SMILES provided for cheminformatics and modelling use.
Catalog No. 50-003-01349 Supplier Medchemexpress LLC Supplier No. HYW01856110MG
Bis(4-octylphenyl)amine (CAS 101-67-7) is a nitrogen-containing organic compound used as a high-temperature antioxidant additive in lubricants and studied as an anthropogenic environmental contaminant. Supplied as a research reagent, it has molecular formula C28H43N and molecular weight 393.65 g·mol⁻¹.
  • Used to evaluate antioxidant performance at elevated temperatures.
  • Relevant for environmental contaminant and fate studies.
  • Available in small, research-scale quantities for analytical work.
  • Supplied with molecular identity information (CAS, formula, mass).
  • SMILES provided for cheminformatics and modelling use.