Learn More
Medchemexpress LLC Bis(4-octylphenyl)amine | 101-67-7 | MFCD00048942 | 393.65 g·mol⁻¹ | C28H43N | 5 MG
SDP
Catalog No. 5000300848
Encompass_Preferred
Bis(4-octylphenyl)amine is an aromatic tertiary amine used as a high-temperature antioxidant additive in lubricants and as an analytical reference standard for environmental and materials testing. It is typically used in formulation research, analytical method development, and studies of anthropogenic contaminants.
  • Acts as a high-temperature antioxidant additive for lubricant formulations.
  • Useful as an analytical reference standard for environmental and materials testing.
  • Hydrophobic aromatic structure suitable for nonpolar matrices.
  • Molecular formula C28H43N, molecular weight 393.65 g·mol⁻¹.
  • Supplied as a research-grade chemical for laboratory use.
Catalog No. 50-003-00848 Supplier Medchemexpress LLC Supplier No. HYW0185615MG
Bis(4-octylphenyl)amine is an aromatic tertiary amine used as a high-temperature antioxidant additive in lubricants and as an analytical reference standard for environmental and materials testing. It is typically used in formulation research, analytical method development, and studies of anthropogenic contaminants.
  • Acts as a high-temperature antioxidant additive for lubricant formulations.
  • Useful as an analytical reference standard for environmental and materials testing.
  • Hydrophobic aromatic structure suitable for nonpolar matrices.
  • Molecular formula C28H43N, molecular weight 393.65 g·mol⁻¹.
  • Supplied as a research-grade chemical for laboratory use.