Learn More
Medchemexpress LLC Bis(4-octylphenyl)amine | 101-67-7 | MFCD00048942 | 85.0% | 393.65 g/mol | C28H43N | 1 MG
SDP
Catalog No. 5000301066
Encompass_Preferred
Bis(4-octylphenyl)amine is an aromatic, nitrogen-containing organic compound (C28H43N, MW 393.65 g/mol) described as an anthropogenic contaminant and used as a high-temperature antioxidant additive in lubricants. It is supplied in small, precise quantities for laboratory and research applications.
  • Acts as a high-temperature antioxidant additive in lubricants.
  • Aromatic amine structure suitable for environmental and materials research.
  • Supplied in defined, small-scale quantities for laboratory use.
  • Well-characterized chemical composition (C28H43N) and molecular weight for analytical consistency.
  • Useful as a reagent or reference in assay development and analytical studies.
Catalog No. 50-003-01066 Supplier Medchemexpress LLC Supplier No. HYW0185611MG
Bis(4-octylphenyl)amine is an aromatic, nitrogen-containing organic compound (C28H43N, MW 393.65 g/mol) described as an anthropogenic contaminant and used as a high-temperature antioxidant additive in lubricants. It is supplied in small, precise quantities for laboratory and research applications.
  • Acts as a high-temperature antioxidant additive in lubricants.
  • Aromatic amine structure suitable for environmental and materials research.
  • Supplied in defined, small-scale quantities for laboratory use.
  • Well-characterized chemical composition (C28H43N) and molecular weight for analytical consistency.
  • Useful as a reagent or reference in assay development and analytical studies.