Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-tert-butylphenyl)amine 90.0+%, TCI America™
SDP

Catalog No. B2803500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2803500G 500 g
B280325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2803500G Supplier TCI America Supplier No. B2803500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4627-22-9
Molecular Formula C20H27N
Molecular Weight (g/mol) 281.443
InChI Key OPEKHRGERHDLRK-UHFFFAOYSA-N
PubChem CID 458684
IUPAC Name 4-tert-butyl-N-(4-tert-butylphenyl)aniline
SMILES CC(C)(C)C1=CC=C(C=C1)NC2=CC=C(C=C2)C(C)(C)C

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 195°C
Quantity 500 g
Synonym 4,4′-Di-tert-butyldiphenylamine
Formula Weight 281.44
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-tert-butylphenyl)amine
EINECSNumber (3)-0135
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.