Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-tert-butylphenyl)iodonium Hexafluorophosphate 98.0+%, TCI America™
SDP

Catalog No. B23805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23805G 5 g
B23801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23805G Supplier TCI America Supplier No. B23805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61358-25-6
Molecular Formula C20H26F6IP
Molecular Weight (g/mol) 538.30
MDL Number MFCD06797070
InChI Key CJLLNCQWBHTSCB-UHFFFAOYSA-N
PubChem CID 13906852
IUPAC Name bis(4-tert-butylphenyl)iodanium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1

Specifications

Melting Point 175°C
Color White
Quantity 5 g
Formula Weight 538.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-tert-butylphenyl)iodonium Hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.