Learn More
Strem, An Ascensus Company CAS# 57731-40-5. 5g. Bis(mu-dimethylamino)tetrakis(dimethylamino)digallium, 98%. MFCD11973803
SDP
Supplier:  Strem, An Ascensus Company 3120305G
Encompass
CAS# 57731-40-5. 5g. Bis(mu-dimethylamino)tetrakis(dimethylamino)digallium, 98%. MFCD11973803. Molecular Weight: 403.90. Molecular Formula: C12H36Ga2N6. Color/form: white xtl. Strem# 31-2030. http://www.strem.com/catalog/v/31-2030/
Catalog No. 50-901-11405