Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(6-hydroxy-2-naphthyl) Disulfide 95.0+%, TCI America™
SDP

Catalog No. D02441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D02441G Supplier TCI America Supplier No. D02441G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6088-51-3
Molecular Formula C20H14O2S2
Molecular Weight (g/mol) 350.45
MDL Number MFCD00004083
InChI Key AHXGXXJEEHFHDK-UHFFFAOYSA-N
Synonym DDD, 6,6′C-Dihydroxy-2,2′C-dinaphthyl Disulfide, 6-Hydroxy-2-naphthyl Disulfide
PubChem CID 22463
IUPAC Name 6-[(6-hydroxynaphthalen-2-yl)disulfanyl]naphthalen-2-ol
SMILES C1=CC2=C(C=CC(=C2)SSC3=CC4=C(C=C3)C=C(C=C4)O)C=C1O

Specifications

Melting Point 226°C
Quantity 1 g
Formula Weight 350.45
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(6-hydroxy-2-naphthyl) Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.