Learn More
Medchemexpress LLC Bis(7-methyl-1-octyl) Phthalate-3,4,5,6-d4 | 1332965-90-8 | C26H38D4O4 | 1 MG
SDP
Catalog No. 5000375652
Encompass_Preferred
Bis(7-methyl-1-octyl) Phthalate-3,4,5,6-d4 is a deuterium-labeled compound. Deuterium, a stable heavy isotope of hydrogen, can be incorporated into drug molecules to serve as tracers for quantitation during drug development. This deuteration has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Used as a tracer for quantitation in drug development
  • May affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-75652 Supplier Medchemexpress LLC Supplier No. HY150855S1MG
Bis(7-methyl-1-octyl) Phthalate-3,4,5,6-d4 is a deuterium-labeled compound. Deuterium, a stable heavy isotope of hydrogen, can be incorporated into drug molecules to serve as tracers for quantitation during drug development. This deuteration has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Used as a tracer for quantitation in drug development
  • May affect pharmacokinetic and metabolic profiles of drugs