Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(Acetonitrile)(1,5-Cyclooctadiene)Rhodium(I)Tetrafluoroborate

Catalog No. 6403601G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-036-01G 1 g
64-036-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 64-036-01G Supplier Sigma Aldrich Supplier No. 6403601G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I)tetrafluoroborate [Rh(cod)(MeCN)2]BF4 may be used as a catalyst for the rhodium(I)-catalyzed silylation of aryl iodides and bromides with tetraethoxysilanes to form aryl silanes such as (4-bromophenyl)triethoxysilane and 4-(triethoxysilyl)acetophenone.

Specifications

CAS 32679-02-0
Molecular Formula C12H18BF4N2Rh
Linear Formula C12H18BF4N2Rh
MDL Number MFCD06798085
Quantity 1 g
Molecular Weight (g/mol) 379.99 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.