Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis(acetonitrile)palladium(II) dichloride | 14592-56-4 | 259.43 | 500 MG
SDP

Catalog No. 5000447521
Encompass_Preferred

Bis(acetonitrile)palladium(II) dichloride is a biochemical reagent utilized in life science research. It presents as a solid with a light yellow to orange hue and has the chemical formula C4H6Cl2N2Pd.

  • Biochemical reagent for life science research
  • Solid with light yellow to orange color
  • Chemical formula: C4H6Cl2N2Pd
  • Shipping at room temperature in continental US
  • Storage recommendations: 4°C, protected from light, stored under nitrogen
  • Solution storage: -80°C for 6 months or -20°C for 1 month (protect from light, stored under nitrogen)

Catalog No. 50-004-47521 Supplier Medchemexpress LLC Supplier No. HYW002087500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis(acetonitrile)palladium(II) dichloride is a biochemical reagent utilized in life science research. It presents as a solid with a light yellow to orange hue and has the chemical formula C4H6Cl2N2Pd.

  • Biochemical reagent for life science research
  • Solid with light yellow to orange color
  • Chemical formula: C4H6Cl2N2Pd
  • Shipping at room temperature in continental US
  • Storage recommendations: 4°C, protected from light, stored under nitrogen
  • Solution storage: -80°C for 6 months or -20°C for 1 month (protect from light, stored under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.