Learn More
Alkali Scientific bis Acrylamide, 2% Solution, 500 Milliliters
SDP
Catalog No. 5000660700
Encompass_Preferred
Bis Acrylamide 2% Solution (500mL) is a ready-to-use liquid reagent used in gel preparation for electrophoresis. This solution simplifies the process of preparing polyacrylamide gels by providing a pre-dissolved mixture of bis acrylamide. It is commonly used in protein and nucleic acid electrophoresis, where precise separation of biomolecules based on size and charge is required. The 2% concentration of bis acrylamide in this solution ensures the gel matrix forms properly when combined with acrylamide, providing the desired stability and clarity for electrophoresis. Due to the neurotoxic properties of acrylamide, it is important to follow all safety guidelines when handling this chemical.
Catalog No. 50-006-60700 Supplier Alkali Scientific Supplier No. BIS004CP
Add to Cart
Add to Cart
Bis Acrylamide 2% Solution (500mL) is a ready-to-use liquid reagent used in gel preparation for electrophoresis. This solution simplifies the process of preparing polyacrylamide gels by providing a pre-dissolved mixture of bis acrylamide. It is commonly used in protein and nucleic acid electrophoresis, where precise separation of biomolecules based on size and charge is required. The 2% concentration of bis acrylamide in this solution ensures the gel matrix forms properly when combined with acrylamide, providing the desired stability and clarity for electrophoresis. Due to the neurotoxic properties of acrylamide, it is important to follow all safety guidelines when handling this chemical.