Learn More
Alkali Scientific bis Acrylamide, 500 G
Supplier: Alkali Scientific BIS005CP
Bis Acrylamide, 500 G is a high-purity reagent used in the preparation of polyacrylamide gels for electrophoresis, gel filtration, and other laboratory techniques. Bis Acrylamide is a crosslinking agent that, when combined with acrylamide, forms polyacrylamide gels essential for protein separation and molecular analysis. This 500g powder form allows for the preparation of large quantities of gel, making it ideal for high-throughput research applications. The reagent ensures consistent gel properties, contributing to reproducibility in gel formation and optimal separation of proteins, nucleic acids, and other biomolecules.