Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis-acrylamide (Electrophoresis), Fisher BioReagents

Catalog No. p-24691 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP171-25 25 g Poly Bottle
BP171-100 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BP171-25
This item is not returnable. View return policy
  • Bis-acrylamide is a cross-linking agent used in the preparation of polyacrylamide gels for electrophoresis
  • RNase, DNase, and protease free
  • Specified and tested for use in electrophoresis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym N, N«–Methylene-bis-acrylamide
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Absorbance 0.2 AU max., 0.2 max. (1% solution) at 290nm
Physical Form Solid
Melting Point 300°C
Color White
pH 5
Quantity 25 g
DNase DNase free
Packaging Poly Bottle
Assay Percent Range ≥95 %
Linear Formula CH2CHCONHCH2NHCOCHCH2
Formula Weight 154.17
Percent Purity ≥95%
Grade Electrophoresis
Chemical Name or Material Bis-Acrylamide
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not induce vomiting. Call a physician or Poison Control Center immediately. . .

NFPA

Health:2
Flammability:1
Instability:1
Recommended Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.