Learn More
Alkali Scientific bis Acrylamide (N,N′ Methylenebisacrylamide), 1 Kg
SDP
Catalog No. 5000660215
Encompass_Preferred
Bis Acrylamide (N,N′ Methylenebisacrylamide) 1kg powder is used for preparing polyacrylamide gels essential in electrophoresis. This chemical is combined with acrylamide to crosslink the gel matrix, allowing for the separation of proteins and nucleic acids during electrophoresis based on their size and charge. The high concentration of bis acrylamide in this 1kg package makes it suitable for laboratories that require larger quantities for frequent gel preparations. Bis acrylamide is critical for maintaining gel stability and achieving precise separation of biomolecules in various molecular biology and biochemistry applications. As with all acrylamide-related chemicals, proper handling and storage are necessary due to potential health risks.
Catalog No. 50-006-60215 Supplier Alkali Scientific Supplier No. BIS002CP
Bis Acrylamide (N,N′ Methylenebisacrylamide) 1kg powder is used for preparing polyacrylamide gels essential in electrophoresis. This chemical is combined with acrylamide to crosslink the gel matrix, allowing for the separation of proteins and nucleic acids during electrophoresis based on their size and charge. The high concentration of bis acrylamide in this 1kg package makes it suitable for laboratories that require larger quantities for frequent gel preparations. Bis acrylamide is critical for maintaining gel stability and achieving precise separation of biomolecules in various molecular biology and biochemistry applications. As with all acrylamide-related chemicals, proper handling and storage are necessary due to potential health risks.