Learn More
Hoefer, Inc 30% Bis-Acrylamide Solution 37.5:1, 500 ml
SDP
Catalog No. NC2743283
Encompass_Preferred
Bis-Acrylamide SolutionUltra Pure Molecular Biology GradeAcrylic Acid: <0.001%Conductivity (2%, Water, µmho) : <5pH (1%, 0.1 M NaCl @ 25°C) : 5RNase/DNase/Protease: None Detected
Catalog No. NC2743283 Supplier Hoefer, Inc Supplier No. GR337500
Bis-Acrylamide SolutionUltra Pure Molecular Biology GradeAcrylic Acid: <0.001%Conductivity (2%, Water, µmho) : <5pH (1%, 0.1 M NaCl @ 25°C) : 5RNase/DNase/Protease: None Detected