Learn More
Medchemexpress LLC Bis-acrylate-PEG6 100mg
SDP
Catalog No. 5000204757
Encompass_Preferred
Bis-acrylate-PEG6 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-04757 Supplier Medchemexpress LLC Supplier No. HYW096166100MG
Bis-acrylate-PEG6 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]