Learn More
Medchemexpress LLC Bis-acrylate-PEG6 250mg
SDP
Catalog No. 5000204743
Encompass_Preferred
Bis-acrylate-PEG6 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-04743 Supplier Medchemexpress LLC Supplier No. 250MGHYW096166
Bis-acrylate-PEG6 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]