Learn More
Medchemexpress LLC Bis-acrylate-PEG6 5g
SDP
Catalog No. 5000205999
Encompass_Preferred
Bis-acrylate-PEG6 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05999 Supplier Medchemexpress LLC Supplier No. HYW0961665G
Bis-acrylate-PEG6 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]