Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(aminomethyl)norbornane (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. B385225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B385225G 25 g
B3852500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B385225G Supplier TCI America Supplier No. B385225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56602-77-8
Molecular Formula C9H18N2
Molecular Weight (g/mol) 154.26
MDL Number MFCD19705260
InChI Key RPYFJVIASOJLJS-UHFFFAOYNA-N
Synonym Bicyclo[2.2.1]heptanedimethanamine, Bis(aminomethyl)bicyclo[2.2.1]heptane
PubChem CID 10857171
IUPAC Name 1-[3-(aminomethyl)bicyclo[2.2.1]heptan-2-yl]methanamine
SMILES NCC1C2CCC(C2)C1CN

Specifications

Color Colorless
Quantity 25 g
UN Number 2735
Formula Weight 154.26
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Bis(aminomethyl)norbornane (mixture of isomers)
EINECSNumber (4)-1732
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.