Learn More
Medchemexpress LLC Bis-aminooxy-PEG2 | 98627-71-5 | MFCD00519242 | 95% | C6H16N2O4 | 100 MG
SDP
Catalog No. 5000353062
Encompass_Preferred
Bis-aminooxy-PEG2 is a PEG-based PROTAC linker used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker
  • One ligand is for an E3 ubiquitin ligase
  • The other ligand is for the target protein
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrade target proteins
Catalog No. 50-003-53062 Supplier Medchemexpress LLC Supplier No. HY140408100MG
Bis-aminooxy-PEG2 is a PEG-based PROTAC linker used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker
  • One ligand is for an E3 ubiquitin ligase
  • The other ligand is for the target protein
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrade target proteins