Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-aminooxy-PEG3 | 98627-70-4 | 98.0% | C8H20N2O5 | 50 MG
SDP

Catalog No. 5000357008
Encompass_Preferred

Bis-aminooxy-PEG3 (CAS: 98627-70-4) is a chemical compound with the molecular formula C8H20N2O5 and a molecular weight of 224.25. It is typically a colorless to light yellow liquid. This product is intended for research use only.

  • Purity of 98.0% by NMR
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Appearance is a colorless to light yellow liquid
  • Chemical structure consistent with 1H NMR and MS analysis

Catalog No. 50-003-57008 Supplier Medchemexpress LLC Supplier No. HY14040950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-aminooxy-PEG3 (CAS: 98627-70-4) is a chemical compound with the molecular formula C8H20N2O5 and a molecular weight of 224.25. It is typically a colorless to light yellow liquid. This product is intended for research use only.

  • Purity of 98.0% by NMR
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Appearance is a colorless to light yellow liquid
  • Chemical structure consistent with 1H NMR and MS analysis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.