Learn More
Medchemexpress LLC Bis-ANS (dipotassium) | 65664-81-5 | 98.2% | 1 MG
SDP
Catalog No. 5000338253
Encompass_Preferred
Bis-ANS dipotassium is a fluorescent probe for hydrophobic proteins. It binds to tubulin with a Kd of 2 μM. It is also a potent biphasic modulator of protein liquid-liquid phase separation (LLPS), promoting LLPS at low concentrations and suppressing it at high concentrations. This compound has been utilized to probe exposed hydrophobic patches in proteins, monitor protein aggregate formation, and modulates the liquid-liquid phase separation of TDP-43 low complexity domain (LCD).
  • Fluorescent probe for hydrophobic proteins
  • Binds to tubulin with a Kd of 2 μM
  • Potent biphasic modulator of protein liquid-liquid phase separation (LLPS)
  • Promotes LLPS at low concentrations
  • Suppresses LLPS at high concentrations
  • Used to probe exposed hydrophobic patches in proteins
  • Monitors protein aggregate formation
  • Modulates the liquid-liquid phase separation of TDP-43 low complexity domain (LCD)
Catalog No. 50-003-38253 Supplier Medchemexpress LLC Supplier No. HY1298111MG
Bis-ANS dipotassium is a fluorescent probe for hydrophobic proteins. It binds to tubulin with a Kd of 2 μM. It is also a potent biphasic modulator of protein liquid-liquid phase separation (LLPS), promoting LLPS at low concentrations and suppressing it at high concentrations. This compound has been utilized to probe exposed hydrophobic patches in proteins, monitor protein aggregate formation, and modulates the liquid-liquid phase separation of TDP-43 low complexity domain (LCD).
  • Fluorescent probe for hydrophobic proteins
  • Binds to tubulin with a Kd of 2 μM
  • Potent biphasic modulator of protein liquid-liquid phase separation (LLPS)
  • Promotes LLPS at low concentrations
  • Suppresses LLPS at high concentrations
  • Used to probe exposed hydrophobic patches in proteins
  • Monitors protein aggregate formation
  • Modulates the liquid-liquid phase separation of TDP-43 low complexity domain (LCD)