Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis-Boc-3.5-diaminobenzoic aci, Advanced ChemTech

Catalog No. 01340874
Click to view available options
Quantity:
1 g
5 g

Catalog No. 01-340-874

Supplier: Advanced Chem Tech BX21105G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133887-83-9
Molecular Formula C17H24N2O6
Molecular Weight (g/mol) 352.39
MDL Number MFCD00270356
InChI Key SSZRVVHPLPWQCW-UHFFFAOYSA-N
IUPAC Name 3,5-bis({[(tert-butoxy)carbonyl]amino})benzoic acid
SMILES CC(C)(C)OC(=O)NC1=CC(=CC(NC(=O)OC(C)(C)C)=C1)C(O)=O

Specifications

Color White to Off-White
Quantity 5 g
Grade Biochemical
Chemical Name or Material Bis-Boc-3.5-diaminobenzoic aci
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.