Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(carbonyldithio)tetrathiafulvalene 95.0+%, TCI America™
SDP

Catalog No. B12721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B12721G Supplier TCI America Supplier No. B12721G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64394-47-4
Molecular Formula C8O2S8
Molecular Weight (g/mol) 384.57
MDL Number MFCD00059965
InChI Key JVSSQMMIVZRMMO-UHFFFAOYSA-N
Synonym 2,2′C-Bis(1,3,4,6-tetrathiapentalene-5-one)
PubChem CID 11429069
IUPAC Name 5-{5-oxo-2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiol-2-ylidene}-2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiol-2-one
SMILES O=C1SC2=C(S1)SC(S2)=C1SC2=C(SC(=O)S2)S1

Specifications

Quantity 1 g
Formula Weight 384.57
Percent Purity ≥95.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Bis(carbonyldithio)tetrathiafulvalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.