Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(Chloromethyl) Ether, >95% min., Ultra Scientific

Catalog No. USRCC006
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RCC-006 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-RCC-006 Supplier Agilent Technologies Supplier No. RCC006
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 542-88-1
Molecular Formula C2H4Cl2O
Molecular Weight (g/mol) 114.953
InChI Key HRQGCQVOJVTVLU-UHFFFAOYSA-N
PubChem CID 10967
ChEBI CHEBI:82270
IUPAC Name chloro(chloromethoxy)methane
SMILES C(OCCl)Cl

Specifications

CAS Max % 100.0
Assay Percent Range >95% min.
Solvent or Matrix Neat
Quantity 100 mg
Chemical Name or Material Bis(Chloromethyl) Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.