Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(chloromethyl) Sulfide 97.0+%, TCI America™
SDP

Catalog No. B08431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08431G 1 g
B08435G 5 g
B084325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B08431G Supplier TCI America Supplier No. B08431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3592-44-7
Molecular Formula C2H4Cl2S
Molecular Weight (g/mol) 131.01
MDL Number MFCD00059474
InChI Key KRIFIIWBVJKVST-UHFFFAOYSA-N
Synonym Chloromethyl Sulfide, Dichlorodimethyl Sulfide, Di(chloromethyl) Sulfide
PubChem CID 565560
IUPAC Name chloro[(chloromethyl)sulfanyl]methane
SMILES ClCSCCl

Specifications

Color Colorless
Boiling Point 156°C
Quantity 1 g
UN Number 1992
Formula Weight 131.01
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(chloromethyl) Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.