Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis(choline)tetrathiomolybdate | 649749-10-0 | 98.0% | 432.56 | 1 ML
SDP

Catalog No. 5000395267
Encompass_Preferred

ATN-224 is an orally active Cu2+/Zn2+-superoxide dismutase 1 (SOD1) inhibitor. It inhibits SOD1 activity in endothelial cells in a dose-dependent manner with an IC50 of 17.5 nM. This product is for research use only and not sold to patients.

  • Orally active SOD1 inhibitor
  • Inhibits SOD1 activity in endothelial cells
  • IC50 of 17.5 nM
  • Supplied as a 10 mM solution in DMSO
  • For research use only
  • Purity of 98.0%
  • Molecular weight of 432.56
  • CAS number 649749-10-0
  • Store at -80°C for 6 months or -20°C for 1 month when in solvent

Catalog No. 50-003-95267 Supplier Medchemexpress LLC Supplier No. HY1607410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

ATN-224 is an orally active Cu2+/Zn2+-superoxide dismutase 1 (SOD1) inhibitor. It inhibits SOD1 activity in endothelial cells in a dose-dependent manner with an IC50 of 17.5 nM. This product is for research use only and not sold to patients.

  • Orally active SOD1 inhibitor
  • Inhibits SOD1 activity in endothelial cells
  • IC50 of 17.5 nM
  • Supplied as a 10 mM solution in DMSO
  • For research use only
  • Purity of 98.0%
  • Molecular weight of 432.56
  • CAS number 649749-10-0
  • Store at -80°C for 6 months or -20°C for 1 month when in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.