Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(Cyclooctadiene)Iridium(I) Tetrakis(3,5-Bis(Trifluoromethyl)Phenyl)Borate

Catalog No. 693774500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-377-4500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-377-4500MG Supplier Sigma Aldrich Supplier No. 693774500MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for:Asymmetric hydrogenation reactionsDirected cyclodehydration reactionDirect prenylation via C-H oxidative addition-allene insertionDouble intramolecular hydroalkoxylationIntramolecular hydroamination reactionsDirected C-H bond cleavageImine vinylation reactionsUmicore Precatalysts for Asymmetric and Cross-Coupling CatalysisProduct of UmicoreAdditional information available at www.pmc.umicore.com

Specifications

CAS 1185-55-3
Density 0.955 g/mL at 25°C
Boiling Point 102°C to 104°C
Molecular Formula C4H12O3Si
Linear Formula CH3Si(OCH3)3
Quantity 500 mg
MDL Number MFCD00008342
Molecular Weight (g/mol) 136.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.