Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)chromium, sublimed, 97+%

Catalog No. AA4317706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4317706 5 g
AA4317703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4317706 Supplier Thermo Scientific Chemicals Supplier No. 04317706
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1271-24-5 | C10H10Cr | 182.19 g/mol

Provides catalyst for ethylene polymerization. A precursor to other chromium(II) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-24-5
Molecular Formula C10H10Cr
Molecular Weight (g/mol) 182.19
MDL Number MFCD00013750
InChI Key TYYBBNOTQFVVKN-UHFFFAOYSA-N
Synonym Chromocene
IUPAC Name Bis(cyclopentadienyl)chromium
SMILES [Cr++].[CH-]1C=CC=C1.[CH-]1C=CC=C1

Specifications

Melting Point 172°C to 173°C
Color Red
Quantity 5 g
UN Number UN3391
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 182.18
Percent Purity ≥97%
Physical Form Crystalline
Chemical Name or Material Bis (cyclopentadienyl)chromium, Sublimed
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC
EINECSNumber 215-036-4
RTECSNumber GB7600000
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.