Learn More
Strem, An Ascensus Company Cobaltocene, min. 98% 1g
SDP
Supplier:  Strem, An Ascensus Company 2704751G
Encompass
CAS# 1277-43-6. 1g. Bis(cyclopentadienyl)cobalt(II), min. 98% (Cobaltocene). MFCD00013749. Molecular Weight: 189.12. Molecular Formula: (C5H5)2Co. Color/form: purplish-black xtl. Strem# 27-0475. http://www.strem.com/catalog/v/27-0475/
Catalog No. 50-901-11171