Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(Cyclopentadienyl)Cobalt(III) Hexafluorophosphate

Catalog No. 18620125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-125ML 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 18-620-125ML Supplier Sigma Aldrich Supplier No. 2798111G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Uses:Cocatalyst for Heck reaction of aryl iodides with alkenes using palladium as catalyst in waterReactant for modification of organometallic electrodes surfaces through cathodic reductionReactant in synthesis and solution self-assembly of side-chain cobaltocenium-containing block copolymers

Specifications

CAS 75-16-1
Density 1.035 g/mL at 25°C
Molecular Formula CH3BrMg
Linear Formula CH3MgBr
MDL Number MFCD00000041
Quantity 200 mL
Synonym Cobaltocenium hexafluorophosphate
Molecular Weight (g/mol) 119.24 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.