Learn More
Medchemexpress LLC Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate | 12427-42-8 | MFCD00011874 | 334.08 g·mol⁻¹ | C10H10CoF6P | 500 MG
SDP
Catalog No. 5000299307
Encompass_Preferred
Cobaltocene hexafluorophosphate is the hexafluorophosphate salt of the cobaltocenium ion, an organometallic, redox-active reagent used in synthetic, electrochemical, and materials research. It is typically handled as a crystalline salt and used as a cocatalyst, oxidant, or building block in organometallic and materials chemistry.
  • Redox-active organometallic reagent suitable for redox studies.
  • Used as a cocatalyst or oxidizing agent in organic synthesis.
  • Soluble in polar aprotic solvents such as acetone and dimethylformamide.
  • Supplied in multiple pack sizes for research laboratories.
  • Applicable to electrochemistry, materials, and organometallic synthesis.
Catalog No. 50-002-99307 Supplier Medchemexpress LLC Supplier No. HYW034796500MG
Cobaltocene hexafluorophosphate is the hexafluorophosphate salt of the cobaltocenium ion, an organometallic, redox-active reagent used in synthetic, electrochemical, and materials research. It is typically handled as a crystalline salt and used as a cocatalyst, oxidant, or building block in organometallic and materials chemistry.
  • Redox-active organometallic reagent suitable for redox studies.
  • Used as a cocatalyst or oxidizing agent in organic synthesis.
  • Soluble in polar aprotic solvents such as acetone and dimethylformamide.
  • Supplied in multiple pack sizes for research laboratories.
  • Applicable to electrochemistry, materials, and organometallic synthesis.