Learn More
Bis(cyclopentadienyl)cobalt, Thermo Scientific Chemicals
Description
Bis(cyclopentadienyl)cobalt is used to catalyze the preparation of pyridines from alkynes and nitriles. It acts as a polymerization inhibitor of olefins. It is a common one-electron reducing agent in the laboratory. It reacts with carbon monoxide to prepare cobalt(I) derivative, cyclopentadienylcobalt dicarbonyl. It also serves as an internal standard in cyclic voltametry. Further, it finds use in Diels-Alder reactions. In addition to this, it is used as a paint drier and as an oxygen stripping agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Melting Point | 176°C to 180°C (decomposition) |
Quantity | 1 g |
UN Number | UN3181 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 189.12 |
Physical Form | Powder |
Chemical Name or Material | Bis(cyclopentadienyl)cobalt |
Safety and Handling
Hazard Category | H228 |
Hazard Statement | GHS H Statement H228-H341-H317 Flammable solid. Suspected of causing genetic defects. May cause an allergic skin reaction. |
Precautionary Statement | P210-P240-P241-P280-P370+P378q |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S. |
EINECSNumber | 215-061-0 |
RTECSNumber | GG0350000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.