Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)hafnium dichloride

Catalog No. AA3610503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3610503 1 g
AA3610506 5 g
AA3610514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3610503 Supplier Thermo Scientific Chemicals Supplier No. 03610503
Add to Cart

CAS: 12116-66-4 | C10H10Cl2Hf | 379.58 g/mol

Starting material to prepare many kinds of hafnocene(II or IV) derivatives; in combination with AgI salt, is an effective activator for glycosyl fluoride. Pharmaceutical intermediates and alos used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12116-66-4
Molecular Formula C10H10Cl2Hf
Molecular Weight (g/mol) 379.58
MDL Number MFCD00001438,MFCD00001438
InChI Key CSEGCHWAMVIXSA-UHFFFAOYSA-L
Synonym bis cyclopentadienyl hafniumdichloride
PubChem CID 131674741
SMILES [Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1

Specifications

Melting Point 230°C to 233°C
Quantity 1 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Soluble in aromatic solvents,chloroform; slightly soluble in THF,ether. Insolble in hexanes.
IUPAC Name cyclopenta-1,3-diene;hafnium;dichloride
Formula Weight 379.59
Physical Form Crystalline
Chemical Name or Material Bis(cyclopentadienyl)hafnium dichloride
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 235-177-5
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.