Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)iron, 99%, Thermo Scientific™

Catalog No. AAA1411809
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1411809 10 g
AAA1411806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1411809 Supplier Thermo Scientific Chemicals Supplier No. A1411809

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-54-5
Molecular Formula C10H10Fe
Molecular Weight (g/mol) 186.04
MDL Number MFCD00001427
InChI Key DFRHTHSZMBROSH-UHFFFAOYSA-N
Synonym ferrocene, bis cyclopentadienyl iron
PubChem CID 25199998
IUPAC Name Ferrocene
SMILES [Fe].c1cccc1.c1cccc1

Specifications

Melting Point 173°C to 174°C
Boiling Point 249°C
Odor Characteristic
Quantity 10 g
UN Number UN1325
Merck Index 14,4037
Formula Weight 186.04
Percent Purity 99%
Chemical Name or Material Bis(cyclopentadienyl)iron
Hazard Category Danger
Hazard Statement GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.
Precautionary Statement GHS P Statement
P210-P280-P240-P241-P301+P312-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.