Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)magnesium, sublimed, 97%, Thermo Scientific™

Catalog No. AA4343406
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4343406 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4343406 Supplier Thermo Scientific Chemicals Supplier No. 4343406

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Specifications

CAS 1284-72-6
Boiling Point 290°C
Packaging Ampule
Molecular Formula C10H10Mg
Quantity 5g
MDL Number MFCD00134487
UN Number UN3391
Sensitivity Air & Moisture Sensitive
Synonym bis cyclopentadienyl magnesium, magnesium cyclopentadienide, bis cyclopentadienyl magnesium ii, bis cyclopentadienyl magnesium mg elec. gr.
InChI Key WGESBNRUPISICS-UHFFFAOYSA-N
SMILES [Mg++].[CH-]1[CH-][CH-][CH-][CH-]1.c1cc[c-]c1
IUPAC Name magnesium;cyclopenta-1,3-diene
Molecular Weight (g/mol) 154.50
PubChem CID 24942211
Formula Weight 154.49
Percent Purity 97%
Chemical Name or Material Bis (cyclopentadienyl)magnesium, Sublimed
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC
storageNote1
  • Ampouled under argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.