Learn More
Strem, An Ascensus Company Manganocene 98% 1g
SDP
Catalog No. 5090110884
Encompass
CAS# 73138-26-8. 1g. Bis(cyclopentadienyl)manganese, 98+% (Manganocene). MFCD00045468. Molecular Weight: 185.13. Molecular Formula: (C5H5)2Mn. Color/form: brown xtl. Strem# 25-0200. http://www.strem.com/catalog/v/25-0200/
Catalog No. 50-901-10884 Supplier Strem, An Ascensus Company Supplier No. 2502001G
CAS# 73138-26-8. 1g. Bis(cyclopentadienyl)manganese, 98+% (Manganocene). MFCD00045468. Molecular Weight: 185.13. Molecular Formula: (C5H5)2Mn. Color/form: brown xtl. Strem# 25-0200. http://www.strem.com/catalog/v/25-0200/