Learn More
Strem, An Ascensus Company CAS# 1271-28-9. 5g. Bis(cyclopentadienyl)nickel, 99% (Nickelocene). MFCD00001441
SDP
Catalog No. 5090111255
Encompass
CAS# 1271-28-9. 5g. Bis(cyclopentadienyl)nickel, 99% (Nickelocene). MFCD00001441. Molecular Weight: 188.90. Molecular Formula: (C5H5)2Ni. Color/form: dark green xtl. Strem# 28-1301. http://www.strem.com/catalog/v/28-1301/
Catalog No. 50-901-11255 Supplier Strem, An Ascensus Company Supplier No. 2813015G
CAS# 1271-28-9. 5g. Bis(cyclopentadienyl)nickel, 99% (Nickelocene). MFCD00001441. Molecular Weight: 188.90. Molecular Formula: (C5H5)2Ni. Color/form: dark green xtl. Strem# 28-1301. http://www.strem.com/catalog/v/28-1301/