Learn More
Strem, An Ascensus Company CAS# 1291-32-3. 100g. Bis(cyclopentadienyl)zirconium dichloride, 99% (Zirconocene dichloride). MFCD00003726
SDP
Catalog No. 5090115246
Encompass
CAS# 1291-32-3. 100g. Bis(cyclopentadienyl)zirconium dichloride, 99% (Zirconocene dichloride). MFCD00003726. Molecular Weight: 292.32. Molecular Formula: (C5H5)2ZrCl2. Color/form: white to off-white pwdr. Strem# 93-4002. http://www.strem.com/catalog/v/93-4002/
Catalog No. 50-901-15246 Supplier Strem, An Ascensus Company Supplier No. 934002100G
CAS# 1291-32-3. 100g. Bis(cyclopentadienyl)zirconium dichloride, 99% (Zirconocene dichloride). MFCD00003726. Molecular Weight: 292.32. Molecular Formula: (C5H5)2ZrCl2. Color/form: white to off-white pwdr. Strem# 93-4002. http://www.strem.com/catalog/v/93-4002/